Culture Media in Bottles
Lieferant: LIOFILCHEM
495270
LIOF495270EA 156 EUR
LIOF495270
Culture Media in Bottles
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
Buffered NaCl Peptone Solution pH 7.0 is used for dissolving, suspending and diluting test samples.
  • Its composition conforms to recommendations in the Harmonized Pharmacopoeias for the microbiological examination of non sterile products
  • This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics
  • Shelf life two years
The powder is very hygroscopic, store the powder at 10 to 30 °C, in a dry environment, in its original container tightly closed. Store bottles and bags at 10 to 25 °C away from light. Do not use the product beyond its expiry date on the label or if product shows any evidence of contamination or any sign of deterioration.