Antibiotic assay discs, multodisc
Lieferant: LIOFILCHEM
95240 95250 95260 95290 95230
LIOF95240EA 204 EUR
LIOF95240 LIOF95250 LIOF95260 LIOF95290 LIOF95230
Antibiotic assay discs, multodisc
Mikrobiologische Testsysteme Antibiotika-Tests
Multodisc with eight antimicrobial agents on a ring.
  • Each specially for a microorganism
Amikacin (AK), Ampicillin (AMP), Ampicillin-Sulbactam (AMS), Carbenicillin (CAR), Cefoxitin (FOX), Ceftazidime (CAZ), Cefuroxime (CXM), Chloramphenicol (C), Clindamycin (CD), Erythromycin (E), Gentamicin (CN), Levofloxacin (LEV), Linezolid (LNZ), Metronidazole (MTZ), Netilmicin (NET), Nitrofurantoin (F), Norfloxacin (NOR), Penicillin G (P), Piperacillin-Tazobactam (TZP), Rifampicin (RD), Streptomycin (S), Teicoplanin (TEC), Tetracycline (TE), Ticarcillin (TC), Tobramycin (TOB), Trimethoprim-Sulfamethoxazole (SXT), Vancomycin (VA). The numbers shows the concentration in μg.