Water samplers, SP 3
Lieferant: MAXX
10020210 10020050 10020025 10020012G 10020012 1002000 10020410 10020414 10020406
MAXA10020210EA 0 EUR
MAXA10020210 MAXA10020050 MAXA10020025 MAXA10020012G MAXA10020012 MAXA1002000 MAXA10020410 MAXA10020414 MAXA10020406
Water samplers, SP 3
Probenehmer Probenehmer für Wasser
Water samplers with thermostatic control and protective top, which can be opened for connection and maintenance works. Self-contained, automatic cooling and heating of the sample compartment to 4 °C independent of the programmable controller, with protection against overheating and automatic defrost cycle.
  • Selectable multi-language function
  • Distribution system easily changed
  • Waterproof foil keyboard and backlit LCD
Zone 2 protection class II 3G EEX nC/R/L IIB T3.
Standards: ISO 5667