Luminata™ Chemiluminescent ELISA HRP Substrates
Lieferant: Merck
ELLUR0200
ELLUR0100
ELLUF0100
MILFELLUR0200EA
517
EUR
MILFELLUR0200
MILFELLUR0100
MILFELLUF0100
Luminata™ Chemiluminescent ELISA HRP Substrates
Elektrophorese-Reagenzien
Western Blotting Reagents
Chemiluminescent HRP detection reagent for use on immunoblots, Western blotting, dot/slot blotting.
The reagent has been matched to optimise the range and sensitivity of detection using various antibodies in ELISA assays.
Stable for 1 year at 4 °C from date of shipment. Contains hydrogen peroxide. Use in well-ventilated area.