Luminata™ Western HRP Substrates Samples for Antibodies of All Affinities
Lieferant: Merck
WBLUM0100
MILFWBLUM0100EA
367
EUR
MILFWBLUM0100
Luminata™ Western HRP Substrates Samples for Antibodies of All Affinities
Elektrophorese-Reagenzien
Western Blotting Reagents
Western blotting, binding assays, amino acid analysis, N-terminal protein sequencing, dot/slot blotting, glycoprotein visualisation, lipopolysaccharide analysis.
- Detection method: Chemiluminescence
Lieferumfang: Components includes:
Luminata classico western HRP substrate, 100 ml (MILFWBLUC0100)
Luminata crescendo western HRP substrate, 100 ml (MILFWBLUR0100)
Luminata forte western HRP substrate, 100 ml (MILFWBLUF0100)