Enrichment pouches
Lieferant: 3M Food Safety
QEBPW225
MMMPQEBPW225EA 188 EUR
MMMPQEBPW225
Enrichment pouches
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
The 3M™ Enrichment Pouches with 225 ml Buffered Peptone Water Broth are heavyweight, puncture-resistant pouches, ideal for many sample preparation and enrichment procedures.
  • Gamma irradiated, ready-to-use pouch with 225 ml Buffered Peptone Water Broth requires no mixing or sterilising
  • Flat bottom, stand-up pouch ensures stability when adding samples
  • Wire tie holds top open for easy sample addition and closes pouch for transportation
  • Multi-laminate polyester pouch is puncture resistant
  • 2 year shelf life from date of manufacture provides long-term readiness
Designed to meet stringent laboratory testing needs, this pouch has the strength needed to endure sample preparation and handling processes. Pre-measured with 225 ml of enrichment broth, the pouch is ready to use with no mixing required. Just add the sample, homogenise to mix, then place directly into the incubator. The broth and pouch are decontaminated through gamma irradiation to maintain organism viability and long shelf life. The flat bottom allows the pouch to stand on its own and heavy gage wire closure holds the pouch open, allowing the technician to work with both hands. Once the desired sample has been added, the open top is folded over itself and the wire tie bent over the fold to seal. The multi-layer pouch is strong enough to use with a paddle blender to effectively mix the sample with the enrichment solution. This product is made in the USA with globally sourced materials and is ISO 9001:2008 certified for quality assurance. It is intended for lab use only.
Buffered Peptone Water Broth is used as a diluent or for non selective enrichment of microorganisms such as Salmonella species or E. coli from foods and environmental samples prior to selective enrichment and isolation. Microorganisms that may be present in a food product can be sub-lethally injured by food processing techniques and may not be recovered by direct selection techniques. Buffered Peptone Water Broth promotes the recovery of those microorganisms that may be injured.