Anti-Glial Fibrillary Acidic Protein Cocktail (SMI-22) Mouse Monoclonal Antibody [clone: SMI-22]

Lieferant: Merck Millipore (Oncogene)
NE1015-100
NE1015-100EA 529 EUR
NE1015-100
Anti-Glial Fibrillary Acidic Protein Cocktail (SMI-22) Mouse Monoclonal Antibody [clone: SMI-22]
Antikörper
Type: Primary
Antigen: Glial Fibrillary Acidic Protein Cocktail (SMI-22)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR