Anti-Myelin Basic Protein (SMI-99) Mouse Monoclonal Antibody [clone: SMI-99]

Lieferant: Merck Millipore (Oncogene)
NE1019-100
NE1019-100EA 496 EUR
NE1019-100
Anti-Myelin Basic Protein (SMI-99) Mouse Monoclonal Antibody [clone: SMI-99]
Antikörper
Type: Primary
Antigen: Myelin Basic Protein (SMI-99)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR