Anti-CYP19A1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Novus Biologicals
NB100-1596F
NOVUNB100-1596FEA 723 EUR
NOVUNB100-1596F
Anti-CYP19A1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: CYP19A1 (cytochrome P450, family 19, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR