Anti-DSG3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5G11]

Lieferant: Novus Biologicals
NB100-1643PE
NOVUNB100-1643PEEA 679 EUR
NOVUNB100-1643PE
Anti-DSG3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5G11]
Antikörper
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5G11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR