Anti-DSG3 Mouse Monoclonal Antibody [clone: 5G11]

Lieferant: Novus Biologicals
NB100-1643SS NB100-1643
NOVUNB100-1643SSEA 236 EUR
NOVUNB100-1643SS NOVUNB100-1643
Anti-DSG3 Mouse Monoclonal Antibody [clone: 5G11]
Antikörper
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR