Anti-RAD51C Mouse Monoclonal Antibody [clone: 2H11/6] (DyLight® 550)

Lieferant: Novus Biologicals
NB100-177R
NOVUNB100-177REA 650 EUR
NOVUNB100-177R
Anti-RAD51C Mouse Monoclonal Antibody [clone: 2H11/6] (DyLight® 550)
Antikörper
Mouse Monoclonal Rad51C Antibody (2H11/6) [DyLight 550]. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: RAD51C
Clonality: Monoclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR