Anti-IREB2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB100-1798B
NOVUNB100-1798BEA 690 EUR
NOVUNB100-1798B
Anti-IREB2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: IREB2 (iron-responsive element binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR