Anti-DEC1 rabbit polyclonal antibody
Lieferant: Novus Biologicals
NB100-1800
NOVUNB100-1800EA 732 EUR
NOVUNB100-1800
Anti-DEC1 rabbit polyclonal antibody
Antikörper
The DEC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to 43070. This antibody reacts with human, mouse. The DEC1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Simple Western