Anti-Gankyrin Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB100-1834B
NOVUNB100-1834BEA 668 EUR
NOVUNB100-1834B
Anti-Gankyrin Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Gankyrin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR