Anti-SLC1A3 Rabbit Polyclonal Antibody (DyLight® 405)

Lieferant: Novus Biologicals
NB100-1869V
NOVUNB100-1869VEA 660 EUR
NOVUNB100-1869V
Anti-SLC1A3 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: SLC1A3 (solute carrier family 1 (glial high affinity glutamate transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR