Anti-Aldo-keto Reductase 1C3/AKR1C3 Goat Polyclonal Antibody
Lieferant: Novus Biologicals
NB100-1940
NOVUNB100-1940EA 622 EUR
NOVUNB100-1940
Anti-Aldo-keto Reductase 1C3/AKR1C3 Goat Polyclonal Antibody
Antikörper
The Aldo-keto Reductase 1C3 / AKR1C3 Antibody from Novus Biologicals is a goat polyclonal antibody to Aldo-keto Reductase 1C3 / AKR1C3. This antibody reacts with human. The Aldo-keto Reductase 1C3 / AKR1C3 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Peptide
Type: Primary
Antigen: Aldo-keto Reductase 1C3/AKR1C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human