Anti-JARID2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Novus Biologicals
NB100-2214H
NOVUNB100-2214HEA 723 EUR
NOVUNB100-2214H
Anti-JARID2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: JARID2 (jumonji, AT rich interactive domain 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR