Anti-LRP8 Rabbit Polyclonal Antibody (DyLight® 405)

Lieferant: Novus Biologicals
NB100-2216V
NOVUNB100-2216VEA 722 EUR
NOVUNB100-2216V
Anti-LRP8 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: LRP8 (low density lipoprotein receptor-related protein 8, apolipoprotein e receptor)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR