Anti-SEMA3B Rabbit Polyclonal Antibody (DyLight® 650)

Lieferant: Novus Biologicals
NB100-2218C
NOVUNB100-2218CEA 688 EUR
NOVUNB100-2218C
Anti-SEMA3B Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: SEMA3B (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR