Anti-MAP1LC3A Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-2220 NB100-2220SS
NOVUNB100-2220EA 758 EUR
NOVUNB100-2220 NOVUNB100-2220SS
Anti-MAP1LC3A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR