Anti-MAP1LC3A Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Novus Biologicals
NB100-2220F
NOVUNB100-2220FEA 830 EUR
NOVUNB100-2220F
Anti-MAP1LC3A Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR