Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 488)
Lieferant: Novus Biologicals
NB100-2220G
NOVUNB100-2220GEA 803 EUR
NOVUNB100-2220G
Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen