Anti-ZMPSTE24 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-2388
NOVUNB100-2388EA 554 EUR
NOVUNB100-2388
Anti-ZMPSTE24 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZMPSTE24 (zinc metallopeptidase STE24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR