Anti-LOX Rabbit Polyclonal Antibody (DyLight® 550)

Lieferant: Novus Biologicals
NB100-2530R
NOVUNB100-2530REA 690 EUR
NOVUNB100-2530R
Anti-LOX Rabbit Polyclonal Antibody (DyLight® 550)
Antikörper
Rabbit Polyclonal LOX Antibody [DyLight 550]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Chicken, Porcine, Xenopus, Zebrafish.

Recommended Dilutions: Immunohistochemistry (Parafin): 10-20 ?g/ml

Type: Primary
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR