Anti-RNF5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LHM 2]

Lieferant: Novus Biologicals
NB100-2688APC
NOVUNB100-2688APCEA 650 EUR
NOVUNB100-2688APC
Anti-RNF5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LHM 2]
Antikörper
Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: LHM 2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR