Anti-CERK Rabbit Polyclonal Antibody (DyLight® 488)
Lieferant: Novus Biologicals
NB100-2911G
NOVUNB100-2911GEA 668 EUR
NOVUNB100-2911G
Anti-CERK Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: CERK (ceramide kinase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human