Anti-HIF1A Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NB100-428B
NOVUNB100-428BEA 690 EUR
NOVUNB100-428B
Anti-HIF1A Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat