Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 650)
Lieferant: Novus Biologicals
NB100-428C
NOVUNB100-428CEA 690 EUR
NOVUNB100-428C
Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat