Anti-PINK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Novus Biologicals
NB100-493H
NOVUNB100-493HEA 722 EUR
NOVUNB100-493H
Anti-PINK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse