Anti-Cre Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Novus Biologicals
NB100-56133F
NOVUNB100-56133FEA 686 EUR
NOVUNB100-56133F
Anti-Cre Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: Cre
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR