Anti-CARD8 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NB100-56181
NOVUNB100-56181EA 355 EUR
NOVUNB100-56181
Anti-CARD8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CARD8 (caspase recruitment domain family, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human