Anti-XIAP Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56187
NOVUNB100-56187EA 355 EUR
NOVUNB100-56187
Anti-XIAP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR