Anti-AKT3 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NB100-56345
NOVUNB100-56345EA 434 EUR
NOVUNB100-56345
Anti-AKT3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKT3 (v-akt murine thymoma viral oncogene homolog 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat