Anti-HIST2H2AC Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56346
NOVUNB100-56346EA 492 EUR
NOVUNB100-56346
Anti-HIST2H2AC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST2H2AC (histone cluster 2, H2ac)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR