Anti-HIST4H4 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56348
NOVUNB100-56348EA 434 EUR
NOVUNB100-56348
Anti-HIST4H4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR