Anti-CARM1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56374
NOVUNB100-56374EA 494 EUR
NOVUNB100-56374
Anti-CARM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CARM1 (coactivator-associated arginine methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR