Anti-IKB zeta Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56390
NOVUNB100-56390EA 439 EUR
NOVUNB100-56390
Anti-IKB zeta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IKB zeta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR