Anti-IL20RA Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56409
NOVUNB100-56409EA 434 EUR
NOVUNB100-56409
Anti-IL20RA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL20RA (interleukin 20 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR