Anti-VDAC2/3 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56463
NOVUNB100-56463EA 558 EUR
NOVUNB100-56463
Anti-VDAC2/3 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal VDAC2/3 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Feline, Zebrafish.

Type: Primary
Antigen: VDAC2/3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR