Anti-FPR1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-56473
NOVUNB100-56473EA 558 EUR
NOVUNB100-56473
Anti-FPR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FPR1 (formyl peptide receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR