Anti-FOSB Mouse monoclonal antibody unconjugated [clone: 83B1138]

Lieferant: Novus Biologicals
NB100-56530
NOVUNB100-56530EA 596 EUR
NOVUNB100-56530
Anti-FOSB Mouse monoclonal antibody unconjugated [clone: 83B1138]
Antikörper
Type: Primary
Antigen: FOSB (FBJ murine osteosarcoma viral oncogene homolog B)
Clonality: monoclonal
Clone: 83B1138
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR