Anti-NFkB p105/p50 Mouse Monoclonal Antibody [clone: 2J10D7]
Lieferant: Novus Biologicals
NB100-56583
NOVUNB100-56583EA 627 EUR
NOVUNB100-56583
Anti-NFkB p105/p50 Mouse Monoclonal Antibody [clone: 2J10D7]
Antikörper
Mouse Monoclonal NFkB p105/p50 Antibody (2J10D7). Tested Applications: Western Blot. Tested Reactivity: Human.
Type: Primary
Antigen: NFkB p105/p50
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human