Anti-PRNP Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NB100-56605
NOVUNB100-56605EA 434 EUR
NOVUNB100-56605
Anti-PRNP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRNP (prion protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Avian, Bovine, Sheep