Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.1]

Lieferant: Novus Biologicals
NB100-56726
NOVUNB100-56726EA 672 EUR
NOVUNB100-56726
Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.1]
Antikörper
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: TL2.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR