Anti-West Nile Virus Protein E Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NB100-56744
NOVUNB100-56744EA 636 EUR
NOVUNB100-56744
Anti-West Nile Virus Protein E Rabbit Polyclonal Antibody
Antikörper
The West Nile Virus Protein E Antibody from Novus Biologicals is a rabbit polyclonal antibody to West Nile Virus Protein E. This antibody reacts with virus. The West Nile Virus Protein E Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1 ?g/ml
Type: Primary
Antigen: West Nile Virus Protein E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus