Anti-VEGFA Mouse Monoclonal Antibody (DyLight® 755) [clone: VG76e]

Lieferant: Novus Biologicals
NB100-648IR
NOVUNB100-648IREA 806 EUR
NOVUNB100-648IR
Anti-VEGFA Mouse Monoclonal Antibody (DyLight® 755) [clone: VG76e]
Antikörper
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: VG76e
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR