Anti-HIV1 p24 Goat polyclonal antibody

Lieferant: Novus Biologicals
NB100-65514
NOVUNB100-65514EA 610 EUR
NOVUNB100-65514
Anti-HIV1 p24 Goat polyclonal antibody
Antikörper
Goat Polyclonal HIV-1 p24 Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Virus.

Type: Primary
Antigen: HIV1 p24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR