Anti-CTR9 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-68205
NOVUNB100-68205EA 712 EUR
NOVUNB100-68205
Anti-CTR9 Rabbit Polyclonal Antibody
Antikörper
The CTR9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CTR9. This antibody reacts with human, mouse. The CTR9 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:250

Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR