Anti-TUBA Mouse Monoclonal Antibody (DyLight® 650) [clone: DM1A]

Lieferant: Novus Biologicals
NB100-690C
NOVUNB100-690CEA 650 EUR
NOVUNB100-690C
Anti-TUBA Mouse Monoclonal Antibody (DyLight® 650) [clone: DM1A]
Antikörper
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: DM1A
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR